  • Search Results
  • High flow nasal cannula for continuous positive airway pressure weaning in preterm neonates: A single-centre experience.
High flow nasal cannula for continuous positive airway pressure weaning in preterm neonates: A single-centre experience.
Journal of paediatrics and child health (2014-07-22)
Arun Sasi, Atul Malhotra
ABSTRACT
High flow nasal cannula therapy (HFNC) is an emerging method of non-invasive respiratory support therapy for premature infants. Recent evidence around its safety and efficacy for post-extubation respiratory support is encouraging. However, its effect on long-term respiratory outcomes is not known. The aim of this study is to determine the effect of HFNC on respiratory outcomes (chronic lung disease (CLD), need for home oxygen) when used to wean babies from continuous positive airway pressure (CPAP). This retrospective study compared respiratory outcomes in infants born <32 weeks gestation needing any respiratory support at birth in two epochs - epoch I (2004-2007) verus epoch II (2008-2011). HFNC was available to be used as a CPAP weaning modality in epoch II. A total of 1286 infants (epoch I: 597 vs. epoch II: 689), mean (standard deviation) gestation, 28.1 (2.2) versus 28.5 (2.3) weeks were enrolled, with 222 (32%) receiving HFNC in epoch II. Overall median duration of respiratory support increased by 150 h in epoch II (P = 0.03), primarily as HFNC hours. A reduction in duration of invasive ventilation and CPAP along with 6% (P = 0.01) reduction in CLD rates was observed in epoch II. No change in home oxygen therapy rates was noted. In logistic regression, HFNC had no significant effect on rates of CLD or home oxygen. Introduction of HFNC for weaning nasal CPAP seems to have a minimal effect on CLD rates. The effect of HFNC on long-term outcomes needs to be further evaluated.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for TOC analysis
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, HPLC Plus