  • Search Results
  • Design of a surface-immobilized 4-nitrophenol molecularly imprinted polymer via pre-grafting amino functional materials on magnetic nanoparticles.
Design of a surface-immobilized 4-nitrophenol molecularly imprinted polymer via pre-grafting amino functional materials on magnetic nanoparticles.
Journal of chromatography. A (2014-09-07)
Ali Mehdinia, Sahar Dadkhah, Tohid Baradaran Kayyal, Ali Jabbari
ABSTRACT
In order to resolve the low adsorption capacity of the surface molecularly imprinting methods, an approach was developed for the preparation of magnetic imprinted polymers by pre-grafting the amino functional material, 3-aminopropyltriethoxysilane (APTES), on the surface of the silica coated magnetic substrate. APTES was used for amino functionalization of the silica coated Fe3O4 nanoparticles. Amino groups were used for immobilization of the template molecules on the magnetic surface and additionally to react with the terminal vinyl groups of cross-linker, ethylene glycol dimethacrylate (EGDMA), by the Michael addition reaction. In this way, the imprinting sites of the analytes formed on the substrate were increased. The sorbent was synthesized in the presence of 4-nitrophenol (4-NP) and EGDMA as the template and cross-linker, respectively. Different parameters affecting the adsorption, such as pH, desorption solvent type and adsorption time were evaluated and optimized. The prepared magnetic molecularly imprinted polymer (MMIP) showed high adsorption capacity and proper selectivity for the template molecule. The kinetic adsorption curve indicated that 90 min was sufficient to achieve the adsorption equilibrium for MMIP. The maximum adsorption capacity was 129.1 mg g(-1). The experiments exhibited a linear range of 10-3000 μg L(-1) for 4-NP with the correlation coefficient (R(2)) of 0.997. The results of the real sample analysis confirmed the applicability of the proposed MMIP for quantitative analysis of 4-NP in the aqueous samples.
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Supelco
Acetonitrile, analytical standard
Supelco
Phenol, PESTANAL®, analytical standard
Supelco
Toluene, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
4-Nitrophenol solution, certified reference material, 5000 μg/mL in methanol