  • Search Results
  • Simultaneous quantification of vitamin D₃, 25-hydroxyvitamin D₃ and 24,25-dihydroxyvitamin D₃ in human serum by LC-MS/MS.
Simultaneous quantification of vitamin D₃, 25-hydroxyvitamin D₃ and 24,25-dihydroxyvitamin D₃ in human serum by LC-MS/MS.
Scandinavian journal of clinical and laboratory investigation (2014-04-23)
Anders Burild, Henrik L Frandsen, Jette Jakobsen
ABSTRACT
Serum 25-hydroxy-vitamin D is the established biomarker of vitamin D status although serum concentrations of vitamin D and 24,25-dihydroxyvitamin D may also be of interest to understand the in vivo kinetics of serum 25-hydroxyvitamin D. An LC-MS/MS method was developed and validated to quantify vitamin D₃, 25-hydroxyvitamin D₃ and 24,25-dihydroxyvitamin D₃ in serum. After protein precipitation of the serum it was loaded on a HybridSPE column to separate vitamin D metabolites from phospholipids. Vitamin D₃, 25-hydroxyvitamin D₃ and 24,25-dihydroxyvitamin D₃ in the eluate were derivatized by 4-phenyl-1,2,4-triazoline-3,5-dione to improve sensitivity in the following LC-MS/MS analysis. Using only 100 μL serum the limit of quantification was < 0.2 ng/mL for vitamin D₃, 25-hydroxyvitamin D₃ and 24,25-dihydroxyvitamin D₃. The method was validated up to 100 ng/mL (260 nmol/L) for vitamin D₃, up to 100 ng/mL (240 nmol/L) for 24,25-dihydroxyvitamin D₃ and up to 200 ng/mL (499 nmol/L) for 25-hydroxyvitamin D₃. Precision was < 6.5% for vitamin D₃ and 25-hydroxyvitamin D₃ and < 10.2% for 24,25-dihydroxyvitamin D₃. We demonstrate that a method including not only serum 25-hydroxyvitamin D₃ but also vitamin D₃ and 24,25-dihydroxyvitamin D₃ could easily be implemented in most modern biochemical laboratories. The method could be used to study the metabolism of endogenous synthesized vitamin D₃ as well as vitamin D₃ in intervention studies.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Sigma-Aldrich
4-Phenyl-1,2,4-triazoline-3,5-dione, 97%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Supelco
Acetonitrile, analytical standard