  • Search Results
  • The nuclear receptor NR2F2 activates star expression and steroidogenesis in mouse MA-10 and MLTC-1 Leydig cells.
The nuclear receptor NR2F2 activates star expression and steroidogenesis in mouse MA-10 and MLTC-1 Leydig cells.
Biology of reproduction (2014-06-06)
Raifish E Mendoza-Villarroel, Nicholas M Robert, Luc J Martin, Catherine Brousseau, Jacques J Tremblay
ABSTRACT
Testosterone production is dependent on cholesterol transport within the mitochondrial matrix, an essential step mediated by a protein complex containing the steroidogenic acute regulatory (STAR) protein. In steroidogenic Leydig cells, Star expression is hormonally regulated and involves several transcription factors. NR2F2 (COUP-TFII) is an orphan nuclear receptor that plays critical roles in cell differentiation and lineage determination. Conditional NR2F2 knockout prior to puberty leads to male infertility due to insufficient testosterone production, suggesting that NR2F2 could positively regulate steroidogenesis and Star expression. In this study we found that NR2F2 is expressed in the nucleus of some peritubular myoid cells and in interstitial cells, mainly in steroidogenically active adult Leydig cells. In MA-10 and MLTC-1 Leydig cells, small interfering RNA (siRNA)-mediated NR2F2 knockdown reduces basal steroid production without affecting hormone responsiveness. Consistent with this, we found that STAR mRNA and protein levels were reduced in NR2F2-depleted MA-10 and MLTC-1 cells. Transient transfections of Leydig cells revealed that a -986 bp mouse Star promoter construct was activated 3-fold by NR2F2. Using 5' progressive deletion constructs, we mapped the NR2F2-responsive element between -131 and -95 bp. This proximal promoter region contains a previously uncharacterized direct repeat 1 (DR1)-like element to which NR2F2 is recruited and directly binds. Mutations in the DR1-like element that prevent NR2F2 binding severely blunted NR2F2-mediated Star promoter activation. These data identify an essential role for the nuclear receptor NR2F2 as a direct activator of Star gene expression in Leydig cells, and thus in the control of steroid hormone biosynthesis.
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Supelco
Hematoxylin
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Supelco
Forskolin, analytical standard
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
SAFC
Sodium deoxycholate
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
1-Naphthol, PESTANAL®, analytical standard
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material