MilliporeSigma
  • Rapid screening for human-pathogenic Mucorales using rolling circle amplification.
Rapid screening for human-pathogenic Mucorales using rolling circle amplification.
Mycoses (2014-09-05)
S Dolatabadi, M J Najafzadeh, G S de Hoog
ABSTRACT
Mucormycosis has emerged as a relatively common severe mycosis in patients with haematological and allogeneic stem cell transplantation. Source of transmission is from unidentified sources in the environment. Early diagnosis of infection and its source of contamination are paramount for rapid and appropriate therapy. In this study, rolling circle amplification (RCA) is introduced as a sensitive, specific and reproducible isothermal DNA amplification technique for rapid molecular identification of six of the most virulent species (Rhizopus microsporus, R. arrhizus var. arrhizus, R. arrhizus var. delemar, Mucor irregularis, Mucor circinelloides, Lichtheimia ramosa, Lichtheimia corymbifera). DNAs of target species were successfully amplified, with no cross reactivity between species. RCA can be considered as a rapid detection method with high specificity and sensitivity, suitable for large screening.
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Supelco
Aucubin, analytical standard
SAFC
Ethanolamine
Aucubin, primary reference standard
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard