  • Search Results
  • Dynamic growth and shrinkage govern the pH dependence of RecA filament stability.
Dynamic growth and shrinkage govern the pH dependence of RecA filament stability.
PloS one (2015-01-22)
Sung Hyun Kim, Jeehae Park, Chirlmin Joo, Doseok Kim, Taekjip Ha
ABSTRACT
RecA proteins form a long stable filament on a single-stranded DNA and catalyze strand exchange reaction. The stability of RecA filament changes dramatically with pH, yet its detailed mechanism is not known. Here, using a single molecule assay, we determined the binding and dissociation rates of RecA monomers at the filament ends at various pH. The pH-induced rate changes were moderate but occurred in opposite directions for binding and dissociation, resulting in a substantial increase in filament stability in lower pH. The highly charged residues in C-terminal domain do not contribute to the pH dependent stability. The stability enhancement of RecA filament in low pH may help the cell to cope with acidic stress by fine-tuning of the binding and dissociation rates without losing the highly dynamic nature of the filament required for strand exchange.
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard