  • Search Results
  • Sulphation pattern analysis of chemically sulphated polysaccharide LbGp1 from Lycium barbarum by GC-MS.
Sulphation pattern analysis of chemically sulphated polysaccharide LbGp1 from Lycium barbarum by GC-MS.
Food chemistry (2014-10-13)
Yujiao Sun, Wei Sun, Jiatong Guo, Xiuting Hu, Guiping Gong, Linjuan Huang, Hongzhi Cao, Zhongfu Wang
ABSTRACT
The polysaccharide LbGp1 from Lycium barbarum L. was sulphated with sulphur trioxide-pyridine complex in DMF, yielding two sulphated polysaccharides, which were LbGp1-OL-SL with 13.7% sulphate content, and LbGp1-OL-SH with 27.4% sulphate content. The sulphation patterns were analysed using a GC-MS strategy. After a series of sequential chemical derivatisations, the sulphated polysaccharides were converted to partially methylated alditol acetates. All the sulphate groups were replaced by acetyl groups, maintaining the positional information of the original sulphation pattern. The number and position of sulphate substitutions were deduced by comparing the relative molar ratio from methylation analysis between native polysaccharide and sulphated derivatives. In LbGp1-OL-SL, 12.65% of sulphation located on C-5 of Ara, only 0.69% and 0.34% of sulphation occurred on C-4 and C-6 Gal, respectively; while in LbGp1-OL-SH, 24.96% of sulphate groups were found at C-5 of Ara, and 0.40% and 2.02% of sulphate groups were found at C-4 and C-6 Gal, respectively.
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%