  • Search Results
  • Metformin potentiates the anticancer effects of cisplatin under normoxic conditions in vitro.
Metformin potentiates the anticancer effects of cisplatin under normoxic conditions in vitro.
Oncology reports (2014-11-26)
Takashi Uehara, Akira Mitsuhashi, Nobuhide Tsuruoka, Makio Shozu
ABSTRACT
Metformin is a diabetes drug with anticancer properties. Several studies have investigated the effects of metformin combined with chemotherapeutic agents, with controversial results. This study evaluated the efficacy of combined metformin/cisplatin treatment in an endometrial cancer cell line. Ishikawa cells were treated with metformin, cisplatin or both types of treatment. Cell proliferation was evaluated by quantification and colorimetric and thymidine incorporation assays, cell cycle progression was assessed by flow cytometry, and apoptosis by the caspase-3 activity assay. The effects of metformin and cisplatin used in combination were assessed under normoxic (21% O2) and hypoxic (1% O2) conditions. Mitochondrial morphology was examined using the MitoTracker dye, while function was assayed by lactate production. Discrepant results were obtained from the different assays of cell proliferation, with the value obtained from the colorimetric assay being higher than that from cell counts after drug treatment. Combined treatment with metformin (≥2 mM) and cisplatin (1 µM) had additive anti-proliferative effects on cells under normoxic conditions. However, the additive effect of metformin was attenuated under hypoxia. Metformin caused morphological and functional changes in mitochondria, which appeared shortened after exposure to metformin, while the connections between individual mitochondria appeared weaker. Additionally, decreased MitoTracker staining was observed after an 8-h exposure to metformin. The colorimetric assay did not accurately determine the effects of metformin and cisplatin on cell proliferation. The additive effects of metformin on cisplatin-induced inhibition of cell proliferation were attenuated under hypoxic conditions, while metformin compromised mitochondrial structure and function. Additional studies are needed to determine the efficacy of this drug combination in vivo.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Kanamycin sulfate Streptomyces kanamyceticus, powder, γ-irradiated
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade