  • Search Results
  • Efficacy of local delivery of ardipusilloside I using biodegradable implants against cerebral tumor growth.
Efficacy of local delivery of ardipusilloside I using biodegradable implants against cerebral tumor growth.
American journal of cancer research (2015-01-30)
Huan Dang, Ji Wang, Jiang-Xue Cheng, Peng-Yuan Wang, Ying Wang, Li-Fei Cheng, Caigan Du, Xiao-Juan Wang
ABSTRACT
Ardipusilloside I (ADS-I) is a natural compound that can be isolated from the Chinese medicinal herb Ardisiapusilla A.DC, and has been reported to inhibit the growth of glioblastoma cells in cultures. This study was designed to test its efficacy by the delivery using biodegradable implants against glioblastoma in vivo. ADS-I was incorporated into polymer microspheres, which were prepared by a mixture of poly (D, L-lactic acid) and poly (D, L-lactic-co-glycolic acid) polymers and then fabricated into wafers. The anti-glioma activities of ADS-I-loaded wafers were examined by methylthiazol tetrazolium (MTT) assay in cultured rat C6 glioma cells, and by magnetic resonance imaging (MRI) and survival monitoring in C6 glioma-bearing rats. Here, we showed that ADS-I-loaded wafers sustained ADS-I release in vitro for 36 days in Higuchi model of kinetics, and had the same cytotoxic activity as ADS-I in the solution against the growth of C6 glioma cells in cultures. In C6 glioma-bearing rats, ADS-I wafer implants inhibited tumor growth in a dose-dependent matter, and were more effective than the same dosage of ADS-I in the solution. The tumor suppression efficacies of ADS-I wafer implants were positively correlated with an increase in tumor cell apoptosis and prolonged animal survival, and were associated with a decrease in vascular endothelial growth factor, C-reactive protein, tumor necrosis factor-α and interleukin-6, and an increase in interleukin-2 expression. In conclusion, this study demonstrates significant efficacy of local delivery of ADS-I using polymer implants against glioma tumor growth in vivo, suggesting the potential of ADS-I-loaded wafers for glioma treatment.
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Dichloromethane, analytical standard, stabilized
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Dichloromethane, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard