  • Search Results
  • Synthesis, HPLC measurement and bioavailability of the phenolic amide amkamide.
Synthesis, HPLC measurement and bioavailability of the phenolic amide amkamide.
Journal of chromatographic science (2013-05-29)
Jae B Park
ABSTRACT
Amkamide is a phenolic amide whose analogues were recently reported to have potent mitochondria protective activity, although their bioavailability is still unknown. Therefore, in this study, amkamide was synthesized and confirmed by nuclear magnetic resonance, and a high-performance liquid chromatography (HPLC) method was developed for analyzing the amides in biological samples. HPLC separation was performed on a Nova-Pak C18 column using a gradient condition and a coulometric electrochemical detector. The HPLC method was able to produce excellent and reproducible separations of amkamide from other amides (oretamide, becatamide, enferamide and veskamide). All five amides yielded outstanding peak resolutions with detection limits as low as 100 fmol. Therefore, the bioavailability of amkamide together with becatamide was determined in a mouse model. Their plasma concentrations were measured following two oral administrations (2 and 4 mg per 30 g body weight) using the HPLC method. As expected, relatively high amounts of amkamide and becatamide (51-58 µM) were detected at the time after administration when the maximum concentration was reached (approximately 20 and 25 min) at the high dose. The data indicate that HPLC methods with amide standards may be utilized to measure amkamide and its analogues in biological samples with excellent resolution and detection limits.
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, 97%
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Supelco
Vanillic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Tyramine, 98%, FG
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Syringic acid, ≥95% (HPLC)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Benzoic acid, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
Supelco
Syringic acid, analytical standard
Supelco
Tyramine, analytical standard
Protocatechuic acid, primary reference standard
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
3,4-Dihydroxybenzoic acid, analytical standard