  • Search Results
  • In vitro evaluation of the acquisition of resistance, antifungal activity and synergism of Brazilian red propolis with antifungal drugs on Candida spp.
In vitro evaluation of the acquisition of resistance, antifungal activity and synergism of Brazilian red propolis with antifungal drugs on Candida spp.
Journal of applied microbiology (2015-01-08)
B Pippi, A J D Lana, R C Moraes, C M Güez, M Machado, L F S de Oliveira, G Lino von Poser, A M Fuentefria
ABSTRACT
To evaluate the ability of Candida parapsilosis and Candida glabrata to develop phenotypic resistance to a benzophenone enriched fraction obtained from Brazilian red propolis (BZP-BRP) as compared to fluconazole (FLC). To investigate possible synergy between BZP-BRP and FLC and anidulafungin (AND). To analyse the development of resistance, isolates susceptible to these antifungals were cultured in increasing concentrations of FLC and BZP-BRP. The increase in FLC minimum inhibitory concentration for all isolates was evident and the majority developed resistance, whereas none isolated became less susceptible to BZP-BRP. Synergism was investigated by checkerboard method. BZP-BRP demonstrated synergy with FLC and indifference with AND for most isolates. In conclusion, the synergism observed with FLC suggests that BZP-BRP could be a possible therapeutic strategy for the treatment of infections related to FLC-resistant Candida sp. The indiscriminate use of antifungals results in the emergence of drug-resistant strains among previously susceptible populations. BZP-BRP can become an alternative for the treatment of persistent infections caused by Candida sp.
Product Description
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
SAFC
MOPS
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)