  • Search Results
  • Antioxidant activity and protein-polyphenol interactions in a pomegranate (Punica granatum L.) yogurt.
Antioxidant activity and protein-polyphenol interactions in a pomegranate (Punica granatum L.) yogurt.
Journal of agricultural and food chemistry (2014-06-19)
Lorena Trigueros, Aneta Wojdyło, Esther Sendra
ABSTRACT
Pomegranate juice (PGJ) is rich in phenolics which are potent antioxidants but also prone to interact with proteins. A yogurt rich in PGJ (40%) made from arils was elaborated (PGY) to determine the antioxidant activity and to estimate the phenolics-proteins interaction during 28 days of cold storage. Juice, yogurts, and protein-free permeates were analyzed for phenolic composition. Yogurt fermentation modified the anthocyanin profile of the initial PGJ, especially the content in cyanidin-3-O-glucoside. During storage, individual anthocyanin content in PGY decreased but it did not modify yogurt color. The analysis of permeates revealed that the degree of phenol-protein interaction depends on the type of phenolic, ellagic acid and dephinidin-3,5-O-diglucoside being the least bound phenolic compounds. The presence of PGJ in yogurt enhanced radical scavenging performance, whereas all the observed ferric reducing power ability of PGY was strictly due to the PGJ present. The 84.73% of total anthocyanins remained bound to proteins at the first day of storage and 90.06% after 28 days of cold storage, revealing the high affinity of anthocyanins for milk proteins.
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Potassium hydride, in paraffin
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil