  • Search Results
  • Prospective neurobiological effects of the aerial and root extracts and some pure compounds of randomly selected Scorzonera species.
Prospective neurobiological effects of the aerial and root extracts and some pure compounds of randomly selected Scorzonera species.
Pharmaceutical biology (2014-06-13)
F Sezer Senol, Ozlem Bahadir Acikara, Gulcin Saltan Citoglu, Ilkay Erdogan Orhan, Stefano Dall' Acqua, Fevzi Ozgökce
ABSTRACT
Scorzonera L. species (Asteraceae) are edible and as medicinal plants are used for various purposed in folk medicine. The methanol extracts of the aerial parts and roots from 27 Scorzonera taxa were investigated for their possible neurobiological effects. Inhibitory potential of the Scorzonera species was tested against acetylcholinesterase (AChE), butyrylcholinesterase (BChE), and tyrosinase (TYRO) at 100 µg mL(-1) using ELISA microtiter assay. Antioxidant activity of the extracts was tested with radical scavenging activity, metal-chelation capacity, ferric- (FRAP), and phosphomolibdenum-reducing antioxidant power (PRAP) assays. Chlorogenic acid, hyperoside, rutin, and scorzotomentosin-4-O-β-glucoside were also screened in the same manner. Total phenol and flavonoid quantification in the extracts were determined spectrophotometrically. The aerial parts of Scorzonera pisidica (40.25 ± 0.74%) and chlorogenic acid (46.97 ± 0.82%) displayed the highest TYRO inhibition, while the remaining samples showed only trivial inhibition against cholinesterases (2.08 ± 1.35%-25.32 ± 1.37%). The same extract of S. pisidica was revealed to be the most potent in scavenging of all three radicals and FRAP assay. Out of 27 taxa, S. pisidica, in particular, may deserve further investigation for its neuroprotective potential.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
Gallic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)