  • Search Results
  • Method development for the redox speciation analysis of iron by ion chromatography-inductively coupled plasma mass spectrometry and carryover assessment using isotopically labeled analyte analogues.
Method development for the redox speciation analysis of iron by ion chromatography-inductively coupled plasma mass spectrometry and carryover assessment using isotopically labeled analyte analogues.
Journal of chromatography. A (2014-05-14)
Mesay Mulugeta Wolle, Timothy Fahrenholz, G M Mizanur Rahman, Matt Pamuku, H M 'Skip' Kingston, Damien Browne
ABSTRACT
An ion chromatography-inductively coupled plasma mass spectrometry (IC-ICP-MS) method was developed for the redox speciation analysis of iron (Fe) based on in-column complexation of Fe(2+) and Fe(3+) by dipicolinic acid (DPA). The effects of column type, mobile phase composition and molecular ion interference were studied in the method optimization. The carryover of the target species in the IC-ICP-MS method was uniquely and effectively evaluated using isotopically enriched analogues of the analytes ((54)Fe(2+) and (57)Fe(3+)). Standard solutions of the enriched standards were injected into the system following analysis of a sample, and the ratios of the isotopes of iron in the enriched standards were calculated based on the chromatographic peak areas. The concentrations of the analytes carried over from the sample to the enriched standards were determined using the quantitative relationship in isotope dilution mass spectrometry (IDMS). In contrast to the routine way of evaluating carryover effect by injecting a blank solution after sample analysis, the use of isotopically enriched standards identified significant analyte carryover in the present method. Extensive experiments were carried out to systematically identify the source of the carryover and to eliminate the problem; the separation column was found to be the exclusive source. More than 95% of the analyte carryover was eliminated by reducing the length of the column. The detection limit of the IC-ICP-MS method (MDL) for the iron species was 2ngg(-1). The method was used to determine Fe(2+) and Fe(3+) in synthetic aqueous standard solutions and a beverage sample.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Ammonium nitrate, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O
Sigma-Aldrich
Ammonium nitrate, ≥99.0%
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ammonium nitrate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%
Supelco
2,6-Pyridinedicarboxylic acid concentrate, 0.02 M C7H5NO4 in water (0.04N), suitable for ion chromatography, eluent concentrate
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
Ethylene, 99.99%
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)