  • Search Results
  • The association between miR-499a polymorphism and oral squamous cell carcinoma progression.
The association between miR-499a polymorphism and oral squamous cell carcinoma progression.
Oral diseases (2014-04-03)
Y-Y Hou, J-H Lee, H-C Chen, C-M Yang, S-J Huang, H-H Liou, C-C Chi, K-W Tsai, L-P Ger
ABSTRACT
To investigate the association of miR-499a genetic polymorphism with the risk of oral leukoplakia, oral submucous fibrosis (OSF), oral squamous cell carcinoma (OSCC), and clinicopathological outcomes of OSCC. The genotyping of miR-499a T>C (rs3746444) using TagMan assay was conducted in two case-control studies of 1549 subjects. miR-499a-5p and miR-499a-3p were assayed using stem-loop RT-PCR for 63 paired OSCC and adjacent normal tissues. T/C+C/C genotypes [adjusted odds ratio (AOR) 1.84, P = 0.032] and C allelic type (AOR 1.91, P = 0.007) at miR-499a T>C were associated with an increased risk of BQ-related OSF as compared to those with T/T genotype or T allelic type, respectively. Conversely, T/C+C/C genotypes and C allelic type decreased the risk of OSCC, especially for non-BQ-related OSCC (for genotype: AOR 0.49, P = 0.010; for allelic type: AOR 0.50, P = 0.007). Additionally, downregulation of miR-499a-5p was found in OSCC tissues (P = 0.001) and correlated with the TT genotype (P = 0.001). The T/C+C/C genotypes of MiR-499a may contribute to an increased risk of BQ-related OSF, but a decreased risk of OSCC. miR-499a T>C influences the expression levels of miR-499a-5p during the tumorigenesis of OSCC.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O