  • Search Results
  • The isolation of Staphylococcus aureus tea tree oil-reduced susceptibility mutants.
The isolation of Staphylococcus aureus tea tree oil-reduced susceptibility mutants.
Phytotherapy research : PTR (2014-02-13)
Jesus A Cuaron, Santosh Dulal, Peter H Cooke, Nathanial J Torres, John E Gustafson
ABSTRACT
Tea tree oil (TTO)-reduced susceptibility (TTORS) mutants of two Staphylococcus aureus laboratory strains were isolated utilizing TTO gradient plates. Attempts to isolate TTORS mutants employing agar plates containing single TTO concentrations failed. All TTORS mutants demonstrated a small colony variant (SCV) phenotype and produced cells with a smaller diameter, as determined by scanning electron microscopy. The addition of SCV auxotrophic supplements to media did not lead to an increase in TTORS mutant colony size. Revertants were also isolated from the TTORS mutants following growth in drug-free media, and all revertant strains demonstrated phenotypes similar to their respective parent strains. Transmission electron microscopy revealed that an SH1000 TTORS mutant demonstrated a thinner cell wall and novel septal invaginations compared with parent strain SH1000. In addition, comparative genomic sequencing did not reveal any mutations in an SH1000 TTORS mutant previously linked to well-characterized SCV genotypes. This study demonstrates that TTO can select for a unique SCV phenotype.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ethanol, puriss. p.a., ACS reagent, prima fine spirit, without additive, F15 o1, ~96%
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard