  • Search Results
  • Carbon nanotubes induce apoptosis resistance of human lung epithelial cells through FLICE-inhibitory protein.
Carbon nanotubes induce apoptosis resistance of human lung epithelial cells through FLICE-inhibitory protein.
Toxicological sciences : an official journal of the Society of Toxicology (2014-11-22)
Varisa Pongrakhananon, Sudjit Luanpitpong, Todd A Stueckle, Liying Wang, Ubonthip Nimmannit, Yon Rojanasakul
ABSTRACT
Chronic exposure to single-walled carbon nanotubes (SWCNT) has been reported to induce apoptosis resistance of human lung epithelial cells. As resistance to apoptosis is a foundation of neoplastic transformation and cancer development, we evaluated the apoptosis resistance characteristic of the exposed lung cells to understand the pathogenesis mechanism. Passage control and SWCNT-transformed human lung epithelial cells were treated with known inducers of apoptosis via the intrinsic (antimycin A and CDDP) or extrinsic (FasL and TNF-α) pathway and analyzed for apoptosis by DNA fragmentation, annexin-V expression, and caspase activation assays. Whole-genome microarray was performed to aid the analysis of apoptotic gene signaling network. The SWCNT-transformed cells exhibited defective death receptor pathway in association with cellular FLICE-inhibitory protein (c-FLIP) overexpression. Knockdown or chemical inhibition of c-FLIP abrogated the apoptosis resistance of SWCNT-transformed cells. Whole-genome expression signature analysis confirmed these findings. This study is the first to demonstrate carbon nanotube-induced defective death receptor pathway and the role of c-FLIP in the process.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O