  • Search Results
  • Methylation profiling of ductal carcinoma in situ and its relationship to histopathological features.
Methylation profiling of ductal carcinoma in situ and its relationship to histopathological features.
Breast cancer research : BCR (2014-10-22)
Jia-Min B Pang, Siddhartha Deb, Elena A Takano, David J Byrne, Nicholas Jene, Alice Boulghourjian, Anne Holliday, Ewan Millar, C Soon Lee, Sandra A O'Toole, Alexander Dobrovic, Stephen B Fox
ABSTRACT
DNA methylation is a well-studied biomarker in invasive breast cancer, but its role in ductal carcinoma in situ (DCIS) is less well characterized. The aims of this study are to assess the methylation profile in DCIS for a panel of well-characterized genes that are frequently methylated in breast cancer, to investigate the relationship of methylation with pathological features, and to perform a proof-of-principle study to evaluate the practicality of methylation as a biomarker in diagnostic DCIS material. Promoter CpG island methylation for a panel of 11 breast cancer-related genes was performed by methylation-sensitive high resolution melting (MS-HRM). Formalin-fixed, paraffin-embedded (FFPE) biopsies from 72 samples of pure DCIS (DCIS occurring in the absence of synchronous invasive carcinoma), 10 samples of mixed DCIS (DCIS adjacent to invasive carcinoma), and 18 samples of normal breast epithelium adjacent to a DCIS lesion were micro-dissected prior to DNA extraction. Methylation was seen for all the tested genes except BRCA1. RASSF1A was the most frequently methylated gene (90% of DCIS samples) and its methylation was associated with comedo necrosis (p = 0.018). Cluster analysis based on the methylation profile revealed four groups, the highly methylated cluster being significantly associated with high nuclear grade, HER2 amplification, negative estrogen receptor (ER) α status, and negative progesterone receptor (PgR) status, (p = 0.038, p = 0.018, p <0.001, p = 0.001, respectively). Methylation of APC (p = 0.017), CDH13 (p = 0.017), and RARβ (p <0.001) was associated with negative ERα status. Methylation of CDH13 (p <0.001), and RARβ (p = 0.001) was associated with negative PgR status. Methylation of APC (p = 0.013) and CDH13 (p = 0.026) was associated with high nuclear grade. Methylation of CDH13 (p = 0.009), and RARβ (p = 0.042) was associated with HER2-amplification. DNA methylation can be assessed in FFPE-derived samples using suitable methodologies. Methylation of a panel of genes that are known to be methylated in invasive breast cancer was able to classify DCIS into distinct groups and was differentially associated with phenotypic features in DCIS.
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O