  • Search Results
  • Oxidative stress activates AMPK in cultured cells primarily by increasing cellular AMP and/or ADP.
Oxidative stress activates AMPK in cultured cells primarily by increasing cellular AMP and/or ADP.
FEBS letters (2014-08-02)
F Romana Auciello, Fiona A Ross, Naoko Ikematsu, D Grahame Hardie
ABSTRACT
AMPK is known to be activated by oxidative stress. Addition of glucose oxidase to cells generates H2O2 at a constant rate that is opposed by enzymic degradation, providing a good model for physiological oxidative stress. AMPK activation by glucose oxidase correlated with increases in cellular AMP:ATP and was greatly reduced in cells expressing an AMP-insensitive AMPK mutant, although a small degree of activation remained. The effects of increased AMP were partly due to inhibition of Thr172 dephosphorylation. These results suggest that changes in adenine nucleotides, rather than direct oxidative modification, are the major drivers of AMPK activation during oxidative stress.
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
SAFC
MOPS
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis