  • Search Results
  • Methylation of miR-155-3p in mantle cell lymphoma and other non-Hodgkin's lymphomas.
Methylation of miR-155-3p in mantle cell lymphoma and other non-Hodgkin's lymphomas.
Oncotarget (2014-09-12)
Rita Lh Yim, Kwan Yeung Wong, Yok Lam Kwong, Florence Loong, Chung Ying Leung, Raymond Chu, William Wai Lung Lam, Pak Kwan Hui, Raymond Lai, Chor Sang Chim
ABSTRACT
Mantle cell lymphoma (MCL) is an aggressive B-cell non-Hodgkin's lymphoma (NHL). In cancers, tumor suppressive microRNAs may be silenced by DNA hypermethylation. By microRNA profiling of representative EBV-negative MCL cell lines before and after demethylation treatment, miR-155-3p was found significantly restored. Methylation-specific PCR, verified by pyrosequencing, showed complete methylation of miR-155-3p in one MCL cell line (REC-1). 5-aza-2'-deoxycytidine treatment of REC-1 led to demethylation and re-expression of miR-155-3p. Over-expression of miR-155-3p led to increased sub-G1 apoptotic cells and reduced cellular viability, demonstrating its tumor suppressive properties. By luciferase assay, lymphotoxin-beta (LT-β) was validated as a miR-155-3p target. In 31 primary MCL, miR-155-3p was found hypermethylated in 6(19%) cases. To test if methylation of miR-155-3p was MCL-specific, miR-155-3p methylation was tested in an additional 191 B-cell, T-cell and NK-cell NHLs, yielding miR-155-3p methylation in 66(34.6%) including 36(27%) non-MCL B-cell, 24(53%) T-cell and 6(46%) of NK-cell lymphoma. Moreover, in 72 primary NHL samples with RNA, miR-155-3p methylation correlated with miR-155-3p downregulation (p=0.024), and LT-β upregulation (p=0.043). Collectively, miR-155-3p is a potential tumor suppressive microRNA hypermethylated in MCL and other NHL subtypes. As miR-155-3p targets LT-β, which is an upstream activator of the non-canonical NF-kB signaling, miR-155-3p methylation is potentially important in lymphomagenesis.
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Cytosine, ≥99%
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Titanium(IV) chloride solution, 0.09 M in 20% HCl
Sigma-Aldrich
Titanium(IV) chloride solution, 1.0 M in toluene
Supelco
Cytosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard
Fluorouracil impurity C, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Titanium(IV) chloride, ReagentPlus®, 99.9% trace metals basis
Supelco
Uracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Titanium(IV) chloride, ≥99.995% trace metals basis