  • Search Results
  • Diagnostic performance study of an antigen microarray for the detection of antiphospholipid antibodies in human serum.
Diagnostic performance study of an antigen microarray for the detection of antiphospholipid antibodies in human serum.
Clinical chemistry and laboratory medicine (2014-10-03)
Aline R Schindler, Oliver Bleher, Markus A Thaler, Carmen J Kocot, Udo Steigerwald, Günther Proll, Günter Gauglitz, Peter B Luppa
ABSTRACT
The parallelization of clinically relevant antigens in a microarray format is of growing importance due to the ability to measure multiple antigen-antibody interactions. With the development of a microarray for the detection of antiphospholipid antibodies we focussed on one important autoimmune disease that is still diagnostically challenging. Reasons are the heterogeneity of the autoantibodies and the unspecific clinical symptoms. For the covalent immobilization of antigenic structures, glass transducers were coated with 11-aminoundecyltrimethoxysilane (11-AUTMS). In total 35 antiphospholipid syndrome (APS) patients, six patients with lupus erythematosus and 24 healthy controls were investigated on a microarray format using polarized imaging reflectometric interference spectroscopy. The novel surface modification based on the short derivative 11-AUTMS resulted in a selective biosensor allowing a clear differentiation of patient and control samples. It combined proteinogenic as well as phospholipid-derived antigens, namely β2-glycoprotein I (β2-GPI), prothrombin, cardiolipin (CL) and a β2-GPI/CL complex. With optimized regeneration conditions, up to 20 consecutive measurements could be performed on one chip. Sensitivity was determined to be 0.800-0.929, specificity was between 0.733 and 0.969, depending on the respective antigen. Multiplexed determination of serological parameters has a great potential. We have shown that our biosensor is capable of detecting four different APS relevant antibodies in parallel exhibiting a sensitivity and specificity comparable to existing ELISA methods.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%