  • Search Results
  • Collateral sensitivity to cold stress and differential BCL-2 family expression in new daunomycin-resistant lymphoblastoid cell lines.
Collateral sensitivity to cold stress and differential BCL-2 family expression in new daunomycin-resistant lymphoblastoid cell lines.
Experimental cell research (2014-12-17)
David Cerezo, Manuel Cánovas, Pilar García-Peñarrubia, Elena Martín-Orozco
ABSTRACT
The acquisition of a multidrug-resistant (MDR) phenotype by tumor cells is one of the main causes of chemotherapy failure in cancer, and, usually, is due to the increased expression of P-glycoprotein (MDR-1, P-gp, ABCB1), a pump that expels chemotherapeutics from the cell and/or regulates apoptosis. Thus, it is fundamental to find drugs or stress stimuli with a capacity to induce apoptosis in such cells and to identify the mechanisms involved. We address this matter in human cells and establish new daunomycin (DNM)-resistant cell lines (IM-9R) by exposing the parental lymphoblastic cells (IM-9) to increasing doses of the anti-neoplastic drug, daunomycin. The resistance level of IM-9R cell lines, MDR-1 expression and functionality, collateral sensitivity and Bcl-2 and caspases protein expression are analyzed. As a result, we show for the first time that, unlike the parental cells, human lymphoblastic resistant cells exhibit collateral sensitivity to cold stress, confirming that this phenomenon is not exclusive to murine leukemic cells, but a broader one associated with the acquisition of drug resistance. Furthermore, the new resistant cell lines undergo a significant increase in active caspase-3 and -9 levels and drastic changes in Bcl-2 family protein expression during the process of MDR phenotype acquisition.
Product Description
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)