  • Search Results
  • Gallic acid exerts a protective or an anti-proliferative effect on glioma T98G cells via dose-dependent epigenetic regulation mediated by miRNAs.
Gallic acid exerts a protective or an anti-proliferative effect on glioma T98G cells via dose-dependent epigenetic regulation mediated by miRNAs.
International journal of oncology (2015-02-04)
Alessandro Paolini, Valeria Curti, Francesca Pasi, Giuliano Mazzini, Rosanna Nano, Enrica Capelli
ABSTRACT
Glioblastoma multiforme (GBM) is the most malignant primary brain tumor in adulthood, characterized by very high recurrence. Following the limited results for conventional therapies, novel therapeutic agents are under investigation. Among the putative new molecules, gallic acid (GA) represents a promising new anticancer drug. The anticancer effect of this drug has been based on its antioxidant effects. The aim of the present study was to investigate the toxic effects of GA on the T98G human glioblastoma cell line and its capacity to modulate the expression of microRNAs targeting the genes involved in tumor growth and invasion. Cytotoxicity, clonogenic ability and cell migration after GA treatment were tested. Moreover, the expression of miRNAs that target genes for antioxidant mitochondrial enzymes (mir-17-3p), p-21 protein (mir-21-5p) and ATM (mir-421-5p) was determined by qRT-PCR. The results confirmed in the T98G cells the anti-proliferative effect of GA reported for other glioma cell lines and showed that the miRNA expression changes depending on GA concentrations. Different GA concentrations can determine a protective or a toxic effect on tumor cells. Thus, the key for GA to induce a specific anticancer action is to use an optimal concentration that avoids these twin effects.
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
T98G Cell Line human, 92090213
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®
Supelco
Gallic acid, certified reference material, TraceCERT®
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology