  • Search Results
  • Enzymatic cocktails produced by Fusarium graminearum under submerged fermentation using different lignocellulosic biomasses.
Enzymatic cocktails produced by Fusarium graminearum under submerged fermentation using different lignocellulosic biomasses.
FEMS microbiology letters (2014-05-16)
Philippe Debeire, Francois Delalande, Olivier Habrylo, Jean-Marc Jeltsch, Alain Van Dorsselaer, Vincent Phalip
ABSTRACT
Fusarium graminearum was grown on four lignocellulosic substrates (corn cobs, wheat bran, hop cell walls, and birchwood) and glucose as the sole carbon source. Proteomic studies performed on the resulting enzymatic cocktails highlighted a great diversity in the number and type of proteins secreted. The cell wall-degrading enzymes (CWDE) proportion varied greatly from 20% to 69%. Only one of the 57 CWDEs detected in this study was common to the five proteomes. In contrast, 35 CWDEs were specific to one proteome only. The polysaccharide-degradation activities were different depending on the cocktail and the polysaccharide used. F. graminearum strongly modifies the enzymatic cocktail it secretes as a function of the biomass used for growth.
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Xylose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N