  • Search Results
  • Synthesis and biological evaluation of a class of mitochondrially-targeted gadolinium(III) agents.
Synthesis and biological evaluation of a class of mitochondrially-targeted gadolinium(III) agents.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-10-18)
Daniel E Morrison, Jade B Aitken, Martin D de Jonge, Fatiah Issa, Hugh H Harris, Louis M Rendina
ABSTRACT
A structure-activity relationship study of a library of novel bifunctional Gd(III) complexes covalently linked to arylphosphonium cations is reported. Such complexes have been designed for potential application in binary cancer therapies such as neutron capture therapy and photon activation therapy. A positive correlation was found between lipophilicity and cytotoxicity of the complexes. Mitochondria uptake was determined by means of inductively coupled plasma mass spectrometry (ICP-MS), and Gd uptake was determined by means of quantification using synchrotron X-ray fluorescence (XRF) imaging. A negative correlation between lipophilicity and tumour selectivity of the Gd(III) complexes was demonstrated. This study highlights the delicate balance required to minimise in vitro cytotoxicity and optimise in vitro tumour selectivity and mitochondrial localisation for this new class of mitochondrially-targeted binary therapy agents. We also report the highest in vitro tumour selectivity for any Gd agent reported to date, with a T/N (tumour/normal cell) ratio of up to 23.5±6.6.
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Methanol, analytical standard
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%