  • Search Results
  • Hypoglycemic and antihyperglycemic effects of phytopreparations and limonoids from Swietenia humilis.
Hypoglycemic and antihyperglycemic effects of phytopreparations and limonoids from Swietenia humilis.
Phytochemistry (2014-12-24)
Berenice Ovalle-Magallanes, Omar N Medina-Campos, José Pedraza-Chaverri, Rachel Mata
ABSTRACT
An aqueous extract from the seeds of Swietenia humilis (31.6-100mg/kg bw) lowered (p<0.05) blood glucose levels in normal and nicotinamide-streptozotocin (NA-STZ)-induced hyperglycemic mice. Furthermore, when administered to fructose-fed rats with metabolic syndrome, the decoction showed significant antihyperglycemic, hypoglycemic and hypolipidemic effects, as well as an augmentation of hepatic glycogen. Limonoids 2-hydroxy-destigloyl-6-deoxyswietenine acetate (1), humulin B (2), methyl-2-hydroxy-3-β-isobutyroxy-1-oxomeliac-8(30)-enate (3), methyl-2-hydroxy-3-β-tigloyloxy-1-oxomeliac-8(30)-enate (4), humilinolide G (5), humilinolide C (6), methyl-2-hydroxy-3-β-isobutyoyl-8α,30α-epoxy-1-oxo-meliacate (7), and humilinolide H (8), were isolated from a CH2Cl2-MeOH (1:1) extract of the seeds. Compounds 5 and 8 are analogs of compounds 6 and 7. The structure of 5 was unequivocally established by X-ray analysis. When tested in normal and NA-STZ-hyperglycemic mice, compounds 1, 2, and 4 (3.16-31.6 mg/kg bw) decreased glycemia during an oral glucose tolerance test. The present investigation sustains the contemporary popular uses of S. humilis seeds for treating metabolic disorders, including diabetes and dyslipidemia and demonstrates the potential of the mexicanolides as antihyperglycemic agents.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)