MilliporeSigma
  • Facile one-pot synthesis of a aptamer-based organic-silica hybrid monolithic capillary column by "thiol-ene" click chemistry for detection of enantiomers of chemotherapeutic anthracyclines.
Facile one-pot synthesis of a aptamer-based organic-silica hybrid monolithic capillary column by "thiol-ene" click chemistry for detection of enantiomers of chemotherapeutic anthracyclines.
The Analyst (2014-07-30)
Han-Peng Jiang, Jiu-Xia Zhu, Chunyan Peng, Jiajia Gao, Fang Zheng, Yu-Xiu Xiao, Yu-Qi Feng, Bi-Feng Yuan
ABSTRACT
In the current study, we developed a facile strategy for the one-pot synthesis of an aptamer-based organic-silica hybrid monolithic capillary column. A 5'-SH-modified aptamer, specifically targeting doxorubicin, was covalently modified in the hybrid silica monolithic column by a sol-gel method combined with "thiol-ene" click reaction. The prepared monolithic column had good stability and permeability, large specific surface, and showed excellent selectivity towards chemotherapeutic anthracyclines of doxorubicin and epirubicin. In addition, the enantiomers of doxorubicin and epirubicin can be easily separated by aptamer-based affinity monolithic capillary liquid chromatography. Furthermore, doxorubicin and epirubicin spiked in serum and urine were also successfully determined, which suggested that the complex biological matrix had a negligible effect on the detection of doxorubicin and epirubicin. Finally, we quantified the concentration of epirubicin in the serum of breast cancer patients treated with epirubicin by intravenous injection. The developed analytical method is cost-effective and rapid, and biological samples can be directly analyzed without any tedious sample pretreatment, which is extremely useful for monitoring medicines in serum and urine for pharmacokinetic studies.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis