  • Search Results
  • Upregulation of autophagy-related gene-5 (ATG-5) is associated with chemoresistance in human gastric cancer.
Upregulation of autophagy-related gene-5 (ATG-5) is associated with chemoresistance in human gastric cancer.
PloS one (2014-10-21)
Jie Ge, Zihua Chen, Jin Huang, Jinxiang Chen, Weijie Yuan, Zhenghao Deng, Zhikang Chen
ABSTRACT
Autophagy-related gene-5 (ATG-5) is one of the key regulators of autophagic cell death. It has been widely regarded as a protective molecular mechanism for tumor cells during the course of chemotherapy. In the present study, we investigated the expression pattern of ATG-5 and multidrug resistance-associated protein-1 (MRP-1) in 135 gastric cancers (GC) patients who were treated with epirubicin, cisplatin and 5-FU adjuvant chemotherapy (ECF) following surgical resection and explored their potential clinical significance. We found that both ATG-5 (77.78%) and MRP-1 (79.26%) were highly expressed in GC patients. ATG-5 expression was significantly associated with depth of wall invasion, TNM stages and distant metastasis of GC (P<0.05), whereas MRP-1 expression was significantly linked with tumor size, depth of wall invasion, lymph node metastasis, TNM stages and differentiation status (P<0.05). ATG-5 expression was positively correlated with MRP-1 (rp = 0.616, P<0.01). Increased expression of ATG-5 and MPR-1 was significantly correlated with poor overall survival (OS; P<0.01) and disease free survival (DFS; P<0.01) of our GC cohort. Furthermore, we demonstrated that ATG-5 was involved in drug resistant of GC cells, which was mainly through regulating autophagy. Our data suggest that upregulated expression of ATG-5, an important molecular feature of protective autophagy, is associated with chemoresistance in GC. Expression of ATG-5 and MRP-1 may be independent prognostic markers for GC treatment.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Trolamine, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)