MilliporeSigma
  • The NADPH oxidase inhibitor diphenyleneiodonium activates the human TRPA1 nociceptor.
The NADPH oxidase inhibitor diphenyleneiodonium activates the human TRPA1 nociceptor.
American journal of physiology. Cell physiology (2014-06-20)
Hiroka Suzuki, Noriyuki Hatano, Yukiko Muraki, Yuka Itoh, Satoko Kimura, Hidetoshi Hayashi, Kikuo Onozaki, Yoshiaki Ohi, Akira Haji, Katsuhiko Muraki
ABSTRACT
Transient receptor potential ankyrin 1 (TRPA1) is a Ca(2+)-permeable nonselective cation channel expressed in neuronal and nonneuronal cells and plays an important role in acute and inflammatory pain. Here, we show that an NADPH oxidase (NOX) inhibitor, diphenyleneiodonium (DPI), functions as a TRPA1 activator in human embryonic kidney cells expressing human TRPA1 (HEK-TRPA1) and in human fibroblast-like synoviocytes. Application of DPI at 0.03-10 μM induced a Ca(2+) response in HEK-TRPA1 cells in a concentration-dependent manner. The Ca(2+) response was effectively blocked by a selective TRPA1 antagonist, HC-030031 (HC). In contrast, DPI had no effect on HEK cells expressing TRPV1-V4 or TRPM8. Four other NOX inhibitors, apocynin (APO), VAS2870 (VAS), plumbagin, and 2-acetylphenothiazine, also induced a Ca(2+) response in HEK-TRPA1 cells, which was inhibited by pretreatment with HC. In the presence of 5 mM glutathione, the Ca(2+) response to DPI was effectively reduced. Moreover, mutation of cysteine 621 in TRPA1 substantially inhibited the DPI-induced Ca(2+) response, while it did not inhibit the APO- and VAS-induced responses. The channel activity was induced by DPI in excised membrane patches with both outside-out and inside-out configurations. Internal application of neomycin significantly inhibited the DPI-induced inward currents. In inflammatory synoviocytes with TRPA1, DPI evoked a Ca(2+) response that was sensitive to HC. In mice, intraplantar injection of DPI caused a pain-related response which was inhibited by preadministration with HC. Taken together, our findings demonstrate that DPI and other NOX inhibitors activate human TRPA1 without mediating NOX.
Sigma-Aldrich
Plumbagin from Plumbago indica
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Auranofin, ≥98% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
2-Acetylphenothiazine, 95%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
4′-Hydroxy-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Supelco
Allyl isothiocyanate, PESTANAL®, analytical standard
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Capsaicin, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®