Differential proteomic profiling of primary and recurrent chordomas.
Oncology reports (2015-03-05)
Su Chen, Wei Xu, Jian Jiao, Dongjie Jiang, Jian Liu, Tenghui Chen, Zongmiao Wan, Leqin Xu, Zhenhua Zhou, Jianru Xiao
ABSTRACT
Chordomas are locally destructive tumors with high rates of recurrence and a poor prognosis. The mechanisms involved in chordoma recurrence remain largely unknown. In the present study, we examined the proteomic profile of a chordoma primary tumor (CSO) and a recurrent tumor (CSR) through mass spectrum in a chordoma patient who underwent surgery. Bioinformatic analysis of the profile showed that 359 proteins had a significant expression difference and 21 pathways had a striking alteration between the CSO and the CSR. The CSR showed a significant increase in carbohydrate metabolism. Immunohistochemistry (IHC) confirmed that the cancer stem cell marker activated leukocyte cell adhesion molecule (ALCAM or CD166) expression level was higher in the recurrent than that in the primary tumor. The present study analyzed the proteomic profile change between CSO and CSR and identified a new biomarker ALCAM in recurrent chordomas. This finding sheds light on unraveling the pathophysiology of chordoma recurrence and on exploring more effective prognostic biomarkers and targeted therapies against this devastating disease.
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, meets USP testing specifications
Millipore
Urea solution, suitable for microbiology, 40% in H2O
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C