  • Search Results
  • Suggestive evidence on the involvement of polypyrimidine-tract binding protein in regulating alternative splicing of MAP/microtubule affinity-regulating kinase 4 in glioma.
Suggestive evidence on the involvement of polypyrimidine-tract binding protein in regulating alternative splicing of MAP/microtubule affinity-regulating kinase 4 in glioma.
Cancer letters (2015-01-13)
L Fontana, D Rovina, C Novielli, E Maffioli, G Tedeschi, I Magnani, L Larizza
ABSTRACT
MAP/microtubule affinity-regulating kinase 4 (MARK4) is a serine-threonine kinase that phosphorylates microtubule-associated proteins taking part in the regulation of microtubule dynamics. MARK4 is expressed in two spliced isoforms characterized by inclusion (MARK4S) or exclusion (MARK4L) of exon 16. The distinct expression profiles in the central nervous system and their imbalance in gliomas point to roles of MARK4L and MARK4S in cell proliferation and cell differentiation, respectively. Having ruled out mutations and transcription defects, we hypothesized that alterations in the expression of splicing factors may underlie deregulated MARK4 expression in gliomas. Bioinformatic analysis revealed four putative polypyrimidine-tract binding (PTB) protein binding sites in MARK4 introns 15 and 16. Glioma tissues and glioblastoma-derived cancer stem cells showed, compared with normal brain, significant overexpression of PTB, correlated with high MARK4L mRNA expression. Splicing minigene assays revealed a functional intronic splicing silencer in MARK4 intron 15, but mutagenesis of the PTB binding site in this region did not affect minigene splicing, suggesting that PTB may bind to a splicing silencer other than the predicted one and synergistically acting with the other predicted PTB sites. Electrophoretic mobility shift assays coupled with mass spectrometry confirmed binding of PTB to the polypyrimidine tract of intron 15, and thus its involvement in MARK4 alternative splicing. This finding, along with evidence of PTB overexpression in gliomas and glioblastoma-derived cancer stem cells and differentiated progeny, merged in pointing out the involvement of PTB in the switch to MARK4L, consistent with its established role in driving oncogenic splicing in brain tumors.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, analytical standard
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Roche
X-tremeGENE HP DNA Transfection Reagent, High-performance polymer reagent for transfecting many cell lines
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material