  • Search Results
  • Estrogen enhances activity of Wnt signaling during osteogenesis by inducing Fhl1 expression.
Estrogen enhances activity of Wnt signaling during osteogenesis by inducing Fhl1 expression.
Journal of cellular biochemistry (2015-02-14)
Shao-Min Wu, Lan-Hsin Shih, Jing-Yu Lee, Yi-Jun Shen, Hu-Hui Lee
ABSTRACT
Estrogen is a crucial hormone for osteoclast inhibition and for preventing osteoporosis. However, the hormone's role in osteoblast growth and differentiation remains unclear. The complexity of estrogen's role in guiding osteoblast behavior arises partly from crosstalk with other signaling pathways, including Wnt signaling. In this study, we show that the Wnt agonist, LiCl, induced Fhl1 gene expression, which substantially enhanced osteoblast differentiation. Staining with alizarin red revealed that MC3T3-E1 mineralization was enhanced by overexpression of Fhl1. In addition, Fhl1 promoted the expression of the osteogenic markers, Runt-related transcription factor 2 (Runx2), osteocalcin (OCN), and osteopontin (OPN), whereas MC3T3-E1 cells with gene knockdown of Fhl1 exhibited limited mineralization and expression of Runx2, OCN, and OPN. We further demonstrate evidences from quantitative reverse transcription real-time polymerase chain reaction and reporter assay that Fhl1 expression was synergistically stimulated by estrogen (E2) and LiCl, but reduced by the estrogen-receptor inhibitor fulvestrant (ICI 182,780). However, estrogen could not enhance osteogenesis while Fhl1 expression was knocked down. Because estrogen and Wnt signaling frequently interact in developmental processes, we propose that Fhl1 can be an acting molecule mediating both signaling pathways during osteogenesis.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O