  • Search Results
  • Phosphorylation of N-methyl-D-aspartic acid receptor-associated neuronal nitric oxide synthase depends on estrogens and modulates hypothalamic nitric oxide production during the ovarian cycle.
Phosphorylation of N-methyl-D-aspartic acid receptor-associated neuronal nitric oxide synthase depends on estrogens and modulates hypothalamic nitric oxide production during the ovarian cycle.
Endocrinology (2010-04-08)
Jyoti Parkash, Xavier d'Anglemont de Tassigny, Nicole Bellefontaine, Celine Campagne, Danièle Mazure, Valérie Buée-Scherrer, Vincent Prevot
ABSTRACT
Within the preoptic region, nitric oxide (NO) production varies during the ovarian cycle and has the ability to impact hypothalamic reproductive function. One mechanism for the regulation of NO release mediated by estrogens during the estrous cycle includes physical association of the calcium-activated neuronal NO synthase (nNOS) enzyme with the glutamate N-methyl-d-aspartate (NMDA) receptor channels via the postsynaptic density 95 scaffolding protein. Here we demonstrate that endogenous variations in estrogens levels during the estrous cycle also coincide with corresponding changes in the state of nNOS Ser1412 phosphorylation, the level of association of this isoform with the NMDA receptor/postsynaptic density 95 complex at the plasma membrane, and the activity of NO synthase (NOS). Neuronal NOS Ser1412 phosphorylation is maximal on the afternoon of proestrus when both the levels of estrogens and the physical association of nNOS with NMDA receptors are highest. Estradiol mimicked these effects in ovariectomized (OVX) rats. In addition, the catalytic activity of NOS in membrane protein extracts from the preoptic region, i.e. independent of any functional protein-protein interactions or cell-cell signaling, was significantly increased in estradiol-treated OVX rats compared with OVX rats. Finally, lambda phosphatase-mediated nNOS dephosphorylation dramatically impaired NOS activity in preoptic region protein extracts, thus demonstrating the important role of phosphorylation in the regulation of NO production in the preoptic region. Taken together, these results yield new insights into the regulation of neuron-derived NO production by gonadal steroids within the preoptic region and raise the possibility that changes in nNOS phosphorylation during fluctuating physiological conditions may be involved in the hypothalamic control of key neuroendocrine functions, such as reproduction.
Product Description
Millipore
Protein A-Sepharose® from Staphylococcus aureus, lyophilized powder
Sigma-Aldrich
Donkey serum