  • Search Results
  • The role of deoxyribonucleic acid in ribonucleic acid synthesis. III. The inhibition of the enzymatic synthesis of ribonucleic acid and deoxyribonucleic acid by actinomycin D and proflavin.
The role of deoxyribonucleic acid in ribonucleic acid synthesis. III. The inhibition of the enzymatic synthesis of ribonucleic acid and deoxyribonucleic acid by actinomycin D and proflavin.
Proceedings of the National Academy of Sciences of the United States of America (1962-07-15)
J HURWITZ, J J FURTH, M MALAMY, M ALEXANDER
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)