  • Search Results
  • Structural study on syndiotactic polystyrene: 2. Crystal structure of molecular compound with toluene.
Structural study on syndiotactic polystyrene: 2. Crystal structure of molecular compound with toluene.
Polymer (1993)
Chatani Y, et al.