  • Search Results
  • The mechanism of methanol synthesis on copper/zinc oxide/alumina catalysts.
The mechanism of methanol synthesis on copper/zinc oxide/alumina catalysts.
J. Catal. (1988)
Bowker M, et al.