  • Search Results
  • Hydroxylation by electrochemically generated OH. radicals. Mono-and polyhydroxylation of benzoic acid: products and isomer distribution.
Hydroxylation by electrochemically generated OH<SUP>. </SUP>radicals. Mono-and polyhydroxylation of benzoic acid: products and isomer distribution.
The Journal of Physical Chemistry (1995)
Oturan MA and Pinson J.