  • Search Results
  • ADSORPTION OF IRON BY ANION EXCHANGE RESINS FROM HYDROCHLORIC ACID SOLUTIONS1.
ADSORPTION OF IRON BY ANION EXCHANGE RESINS FROM HYDROCHLORIC ACID SOLUTIONS1.
Journal of the American Chemical Society (1950)
Moore GE and Kraus KA.