  • Search Results
  • Alcoholic hydrochloric acid-carmine as a stain for chromosomes in squash preparations.
Alcoholic hydrochloric acid-carmine as a stain for chromosomes in squash preparations.
Stain technology (1963-01-01)
R SNOW
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis