  • Search Results
  • Aldol condensation of acetone over layered double hydroxides of the meixnerite type.
Aldol condensation of acetone over layered double hydroxides of the meixnerite type.
Applied Clay Science (1998)
Tichit D, et al.