  • Search Results
  • An investigation on photocatalytic activities of mixed TiO2-rare earth oxides for the oxidation of acetone in air.
An investigation on photocatalytic activities of mixed TiO<SUB>2</SUB>-rare earth oxides for the oxidation of acetone in air.
Journal of Photochemistry and Photobiology A: Chemistry (1998)
Lin J and Jimmy CY.