  • Search Results
  • Effects of doxorubicin and gemcitabine on the induction of apoptosis in breast cancer cells.
Effects of doxorubicin and gemcitabine on the induction of apoptosis in breast cancer cells.
Oncology reports (2014-10-02)
Rui-Rui Zheng, Wei Hu, Cheng-Guang Sui, Nan Ma, You-Hong Jiang
ABSTRACT
Breast cancer is the most frequently diagnosed cancer and the leading cause of cancer-related mortality in females worldwide. The efficacy of chemotherapeutic drugs on breast cancer is hindered by many factors, including that cancer stem-like side population (SP) cells may contribute to tumorigenesis and resistance to chemotherapy. Doxorubicin and gemcitabine are two of the most effective and widely used chemotherapeutic agents for the treatment of various human malignancies. To the best of our knowledge, the effects of doxorubicin and gemcitabine on breast cancer SP cells are poorly understood. In the present study, we examined the potential roles of doxorubicin and gemcitabine in breast cancer main population (MP) and SP cells, and the mechanisms of doxorubicin and gemcitabine. The results showed that doxorubicin and gemcitabine decreasede proliferation, and increased apoptosis in the MCF7 MP and SP cells in vitro. Furthermore, doxorubicin and gemcitabine inhibited tumor growth and improved the survival rate of mice in vivo. Additionally, the mechanisms of doxorubicin and gemcitabine in MCF7 MP and SP cells were determined.
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Gemcitabine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Anti-CD133 Antibody, clone 13A4, clone 13A4, Chemicon®, from rat
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)