  • Search Results
  • Evaluation of miR-216a and miR-217 as potential biomarkers of acute pancreatic injury in rats and mice.
Evaluation of miR-216a and miR-217 as potential biomarkers of acute pancreatic injury in rats and mice.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2014-07-26)
David Goodwin, Barry Rosenzweig, Jun Zhang, Lin Xu, Sharron Stewart, Karol Thompson, Rodney Rouse
ABSTRACT
Mild injury of the exocrine pancreas is often asymptomatic and can be under- or mis-diagnosed. The pancreas-enriched microRNAs miR-216a and miR-217 were evaluated as potential serum biomarkers of exocrine pancreas injury in rodent models of acute pancreatitis induced by caerulein, l-arginine, and pancreatic duct ligation. Both microRNAs showed time- and dose- relevant responses to pancreatic injury and wider dynamic ranges of response than serum amylase or lipase. Pancreas-selective microRNAs were found to be relatively sensitive serum biomarkers of pancreatic injury in rodents with potentially greater specificity than the current standard assays.
Sigma-Aldrich
DL-Arginine, ≥95% (NT)
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Arginine, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material