  • Search Results
  • BBS4 is necessary for ciliary localization of TrkB receptor and activation by BDNF.
BBS4 is necessary for ciliary localization of TrkB receptor and activation by BDNF.
PloS one (2014-05-29)
Carmen C Leitch, Norann A Zaghloul
ABSTRACT
Primary cilia regulate an expanding list of signaling pathways in many different cell types. It is likely that identification of the full catalog of pathways associated with cilia will be necessary to fully understand their role in regulation of signaling and the implications for diseases associated with their dysfunction, ciliopathies. Bardet-Biedl Syndrome (BBS) is one such ciliopathy which is characterized by a spectrum of phenotypes. These include neural defects such as impaired cognitive development, centrally mediated hyperphagia and peripheral sensory defects. Here we investigate potential defects in a signaling pathway associated with neuronal function, brain derived neurotrophic factor (BDNF) signaling. Upon loss of BBS4 expression in cultured cells, we observed decreased phosphorylation and activation by BDNF of its target receptor, TrkB. Assessment of ciliary localization revealed that, TrkB localized to the axonemes or basal bodies of cilia only in the presence of BDNF. Axonemal localization, specifically, was abrogated with loss of BBS4. Finally, we present evidence that loss of the ciliary axoneme through depletion of KIF3A impedes activation of TrkB. Taken together, these data suggest the possibility of a previously uninvestigated pathway associated with perturbation of ciliary proteins.
Sigma-Aldrich
Anti-phospho-Trk B (pTyr515) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
MOPS
SAFC
BIS-TRIS
SAFC
BIS-TRIS