  • Search Results
  • In vivo antigen-driven plasmablast enrichment in combination with antigen-specific cell sorting to facilitate the isolation of rare monoclonal antibodies from human B cells.
In vivo antigen-driven plasmablast enrichment in combination with antigen-specific cell sorting to facilitate the isolation of rare monoclonal antibodies from human B cells.
Nature protocols (2014-06-06)
Zhonghua Lin, Nancy Y Chiang, Ning Chai, Dhaya Seshasayee, Wyne P Lee, Mercedesz Balazs, Gerald Nakamura, Lee R Swem
ABSTRACT
The ability to rapidly generate large panels of antigen-specific human antibodies in a rodent would enable the efficient discovery of novel therapeutically useful antibodies. We have developed a system wherein human antigen-specific antibody-secreting plasmablasts can be enriched in vivo, in a severe combined immunodeficient (SCID)/beige mouse host. The antigen-specific plasmablasts can then be sorted by flow cytometry, enabling single-cell cloning and expression of fully human immunoglobulin-G. By using this technique, we have generated four broadly reactive anti-influenza A antibodies. Therefore, the method described here is useful for the identification of rare functional antibodies. This protocol takes ∼1 month to complete, from the time of human vaccination to the cloning of heavy- and light-chain genes. For additional small-scale transient expression, purification and binding analysis, the protocol would take an additional month.
Sigma-Aldrich
Bovine Serum Albumin solution, 30% in DPBS, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Tris-EDTA buffer solution, BioUltra, for molecular biology, pH 8.0
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)