  • Search Results
  • Role of 15-F2t-isoprostane in intestinal injury induced by intestinal ischemia/reperfusion in rats.
Role of 15-F2t-isoprostane in intestinal injury induced by intestinal ischemia/reperfusion in rats.
Free radical research (2014-05-23)
S-H Wen, Y-H Ling, W-F Liu, Y-X Qiu, Y-S Li, Y Wu, J-T Shen, Z-Y Xia, K-X Liu
ABSTRACT
15-F2t-isoprostane is not only a specific marker of lipid peroxidation but also demonstrated to have potent bioactivities and can exert deleterious effects via activating thromboxane A2 receptor (TxA2r). We already demonstrated that lipid peroxidation represents a mechanism of intestinal ischemia/reperfusion (I/R) injury. But no studies have focused on 15-F2t-isoprostane production and its biological actions on postischemic intestine during intestinal I/R. This study was carried to investigate whether the mechanism of endogenous 15-F2t-isoprostane action is involved in the pathogenesis of intestinal I/R and administration of synthetic 15-F2t-isoprostane could exacerbate intestinal insult after intestinal I/R in vivo and in vitro. In comparison with that of the sham control, we reported that endogenous 15-F2t-isoprostane was liberated following intestinal I/R injury in rats, and using the TxA2r antagonist SQ29548 resulted in significant intestinal protection, evidenced by reduced lipid peroxidation, inflammation, and alleviated intestinal mucosal microvascular vasoconstriction. Further research found that in vivo administration of synthetic 15-F2t-isoprostane exacerbated intestinal I/R injury by disturbing microvascular perfusion and accumulating anaerobic metabolism. Meanwhile, 15-F2t-isoprostane did not change Hypoxia/Reoxygenation-induced IEC-6 cell viability but aggravated HUVECs cell death in vitro. Collectively, our results showed that locally produced 15-F2t-isoprostane was in proportion to the severity of oxidative stress-induced intestinal injury and its detrimental effects can be attenuated through TxA2r inactivation. Exogenous 15-F2t-isoprostane exacerbated intestinal I/R injury, which may be contributable to its biological actions on endothelium, rather than intestinal epithelium.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%