  • Search Results
  • Overexpression of oxytocin receptors in the hypothalamic PVN increases baroreceptor reflex sensitivity and buffers BP variability in conscious rats.
Overexpression of oxytocin receptors in the hypothalamic PVN increases baroreceptor reflex sensitivity and buffers BP variability in conscious rats.
British journal of pharmacology (2014-05-20)
Maja Lozić, Michael Greenwood, Olivera Šarenac, Andrew Martin, Charles Hindmarch, Tatjana Tasić, Julian Paton, David Murphy, Nina Japundžić-Žigon
ABSTRACT
The paraventricular nucleus (PVN) of the hypothalamus is an important integrative site for neuroendocrine control of the circulation. We investigated the role of oxytocin receptors (OT receptors) in PVN in cardiovascular homeostasis. Experiments were performed in conscious male Wistar rats equipped with a radiotelemetric device. The PVN was unilaterally co-transfected with an adenoviral vector (Ad), engineered to overexpress OT receptors, and an enhanced green fluorescent protein (eGFP) tag. Control groups: PVN was transfected with an Ad expressing eGFP alone or untransfected, sham rats (Wt). Recordings were obtained without and with selective blockade of OT receptors (OTX), during both baseline and stressful conditions. Baroreceptor reflex sensitivity (BRS) and cardiovascular short-term variability were evaluated using the sequence method and spectral methodology respectively. Under baseline conditions, rats overexpressing OT receptors (OTR) exhibited enhanced BRS and reduced BP variability compared to control groups. Exposure to stress increased BP, BP variability and HR in all rats. In control groups, but not in OTR rats, BRS decreased during stress. Pretreatment of OTR rats with OTX reduced BRS and enhanced BP and HR variability under baseline and stressful conditions. Pretreatment of Wt rats with OTX, reduced BRS and increased BP variability under baseline and stressful conditions, but only increased HR variability during stress. OT receptors in PVN are involved in tonic neural control of BRS and cardiovascular short-term variability. The failure of this mechanism could critically contribute to the loss of autonomic control in cardiovascular disease.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)