MilliporeSigma
  • Bone marrow-derived mesenchymal stem cells become antiangiogenic when chondrogenically or osteogenically differentiated: implications for bone and cartilage tissue engineering.
Bone marrow-derived mesenchymal stem cells become antiangiogenic when chondrogenically or osteogenically differentiated: implications for bone and cartilage tissue engineering.
Tissue engineering. Part A (2013-07-31)
Jennifer J Bara, Helen E McCarthy, Emma Humphrey, William E B Johnson, Sally Roberts
ABSTRACT
Osteochondral tissue repair requires formation of vascularized bone and avascular cartilage. Mesenchymal stem cells stimulate angiogenesis both in vitro and in vivo but it is not known if these proangiogenic properties change as a result of chondrogenic or osteogenic differentiation. We investigated the angiogenic/antiangiogenic properties of equine bone marrow-derived mesenchymal stem cells (eBMSCs) before and after differentiation in vitro. Conditioned media from chondrogenic and osteogenic cell pellets and undifferentiated cells was applied to endothelial tube formation assays using Matrigel™. Additionally, the cell secretome was analysed using LC-MS/MS mass spectrometry and screened for angiogenesis and neurogenesis-related factors using protein arrays. Endothelial tube-like formation was supported by conditioned media from undifferentiated eBMSCs. Conversely, chondrogenic and osteogenic conditioned media was antiangiogenic as shown by significantly decreased length of endothelial tube-like structures and degree of branching compared to controls. Undifferentiated cells produced higher levels of angiogenesis-related proteins compared to chondrogenic and osteogenic pellets. In summary, eBMSCs produce an array of angiogenesis-related proteins and support angiogenesis in vitro via a paracrine mechanism. However, when these cells are differentiated chondrogenically or osteogenically, they produce a soluble factor(s) that inhibits angiogenesis. With respect to osteochondral tissue engineering, this may be beneficial for avascular articular cartilage formation but unfavourable for bone formation where a vascularized tissue is desired.
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium thiosulfate solution, 2 g/dL in deionized water
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%