  • Search Results
  • Calreticulin protects rat microvascular endothelial cells against microwave radiation-induced injury by attenuating endoplasmic reticulum stress.
Calreticulin protects rat microvascular endothelial cells against microwave radiation-induced injury by attenuating endoplasmic reticulum stress.
Microcirculation (New York, N.Y. : 1994) (2014-03-05)
Wei-Hong Li, Yu-Zhen Li, Dan-Dan Song, Xiao-Reng Wang, Mi Liu, Xu-Dong Wu, Xiu-Hua Liu
ABSTRACT
This study was designed to evaluate whether exogenous CRT was beneficial for alleviating MR-induced injury by suppressing ER stress in rat MMECs. MMECs were pretreated with CRT (25 pg/mL) for 12 hours, followed by the exposure to 2.856 GHz radiation at a mean power density of 30 mW/cm(2) for six minutes. MR-induced injury in MMECs was evaluated by LDH leakage, apoptosis, and cell viability analysis. The expression of GRP78, CRT, CHOP, Bcl-2, and Bax were examined by Western blot analysis to reflect ER stress response and ER stress-related apoptosis. MR induced marked MMECs injury, as shown by increased LDH leakage and apoptosis rate and decreased cell viability. MR also induced excessive ER stress, characterized by increased expression of GRP78 and CRT, and ER stress-related apoptotic signaling as well, as shown by the upregulation of CHOP and Bax and the downregulation of Bcl-2. Exogenous CRT pretreatment remarkably attenuated MR-induced cell apoptosis and LDH leakage, ER stress, and activation of the ER stress-related apoptotic signaling. Exogenous CRT attenuates MR-induced ER stress-related apoptosis by suppressing CHOP-mediated apoptotic signaling pathways in MMECs.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
HEPES
Supelco
Acrylamide, certified reference material, TraceCERT®
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)