  • Search Results
  • Noninvasive molecular imaging of apoptosis in a mouse model of anthracycline-induced cardiotoxicity.
Noninvasive molecular imaging of apoptosis in a mouse model of anthracycline-induced cardiotoxicity.
Circulation. Cardiovascular imaging (2015-02-07)
Helen Su, Natalia Gorodny, Luis Felipe Gomez, Umesh Gangadharmath, Fanrong Mu, Gang Chen, Joseph C Walsh, Katrin Szardenings, Hartmuth C Kolb, Balaji Tamarappoo
ABSTRACT
Anthracycline-induced cardiotoxicity and myocardial dysfunction may be associated with apoptosis. Caspase 3 catalyzes a terminal step in apoptosis, and its expression may serve as a marker of cardiomyocyte apoptosis. We synthesized 18F-CP18, a caspase-3 substrate and evaluated cardiac 18F-CP18 uptake in a mouse model of anthracycline cardiotoxicity. For 12 weeks, mice were injected with doxorubicin, 3 mg/kg/week, or vehicle (control). Left ventricular fractional shortening was quantified by echocardiography. CP18 uptake after intravenous injection of 250 μCi of 18F-CP18, 24 hours post-doxorubicin treatment was quantified by microPET, autoradiography, and gamma counting. Apoptosis was assessed by enzymatic assay of myocardial caspase 3 and TUNEL staining of tissue sections. Compared with controls, at 6 and 12 weeks of doxorubicin treatment, fractional shortening was reduced (20.7%±2.5% versus 31%±3.5%, P=0.010; and 20.3%±3.1% versus 32.4%±2.1%, P=0.011). Doxorubicin treatment was associated with increased 18F-CP18 uptake in %ID/g by gamma counting from 0.36±0.01 (week 1) to 0.78±0.01 (week 12), P=0.003. A similar increase in 18F-CP18 uptake was observed by microPET (0.41±0.04 versus 0.73±0.1, P=0.014) and autoradiography (1.1±0.3 versus 2.8±0.2 P=0.001). Caspase 3 enzymatic activity and apoptosis by TUNEL staining were also increased after 12 weeks of doxorubicin compared with weeks 1 and 3. CP18 uptake in controls was relatively unchanged at weeks 1, 3, and 12. In a mouse model of cardiotoxicity, doxorubicin treatment is associated with increased myocardial caspase 3 expression and an increase in CP18 uptake. 18F-CP18 may be useful for detection of anthracycline-induced myocardial apoptosis.
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard